Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(chloromethyl)tetramethyldisiloxane 98.0+%, TCI America™
SDP

Catalog No. B097825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B097825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B097825ML Supplier TCI America Supplier No. B097825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2362-10-9
Molecular Formula C6H16Cl2OSi2
Molecular Weight (g/mol) 231.26
MDL Number MFCD00000877
InChI Key NBGGEWGFZUDQKZ-UHFFFAOYSA-N
Synonym 1,3-bis chloromethyl-1,1,3,3-tetramethyldisiloxane, 1,3-bis chloromethyl tetramethyldisiloxane, bis chloromethyl tetramethyldisiloxane, disiloxane, 1,3-bis chloromethyl-1,1,3,3-tetramethyl, tetramethylbis chloromethyl disiloxane, tetramethyl-1,3-bis chloromethyl disiloxane, chloromethyl chloromethyl dimethylsilyl oxy dimethylsilane, disiloxane,3-bis chloromethyl-1,1,3,3-tetramethyl, pubchem13666, acmc-2097an
PubChem CID 75385
IUPAC Name chloromethyl-[chloromethyl(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(CCl)O[Si](C)(C)CCl

Specifications

Color Yellow
Boiling Point 204°C
Quantity 25 mL
Formula Weight 231.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(chloromethyl)tetramethyldisiloxane
EINECSNumber (7)-0500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.