Learn More
Strem, An Ascensus Company CAS# 286014-38-8. 500mg. 1,3-Bis(cyclohexyl)imidazolium tetrafluoroborate, min. 97% ICyHBF4. MFCD04973310
SDP
Catalog No. 501385338
Encompass
CAS# 286014-38-8. 500mg. 1,3-Bis(cyclohexyl)imidazolium tetrafluoroborate, min. 97% ICyHBF4. MFCD04973310. Molecular Weight: 320.18. Molecular Formula: [C15H25N2]+BF4ae. Color/form: white to cream-colored solid Strem# 07-0597. http://www.strem.com/catalog/v/07-0597/
Catalog No. 50-138-5338 Supplier Strem, An Ascensus Company Supplier No. 070597500MG
CAS# 286014-38-8. 500mg. 1,3-Bis(cyclohexyl)imidazolium tetrafluoroborate, min. 97% ICyHBF4. MFCD04973310. Molecular Weight: 320.18. Molecular Formula: [C15H25N2]+BF4ae. Color/form: white to cream-colored solid Strem# 07-0597. http://www.strem.com/catalog/v/07-0597/