Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(dicyanomethylidene)indan 98.0+%, TCI America™
SDP

Catalog No. B14165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14165G Supplier TCI America Supplier No. B14165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38172-19-9
Molecular Formula C15H6N4
Molecular Weight (g/mol) 242.241
MDL Number MFCD00142514
InChI Key HBZYYOYCJQHAEL-UHFFFAOYSA-N
Synonym 1,3-Bis(dicyanomethylene)indan
PubChem CID 3360610
IUPAC Name 2-[3-(dicyanomethylidene)inden-1-ylidene]propanedinitrile
SMILES C1C(=C(C#N)C#N)C2=CC=CC=C2C1=C(C#N)C#N

Specifications

Quantity 5 g
UN Number 3439
Formula Weight 242.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(dicyanomethylidene)indan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.