Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Bis(Dicyclohexylphosphino)Propane Bis(Tetrafluoroborate)

Catalog No. 703028500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-302-8500MG 500 mg
70-302-82G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-302-8500MG Supplier Sigma Aldrich Supplier No. 703028500MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Bis(Dicyclohexylphosphino)Propane Bis(Tetrafluoroborate)

Specifications

CAS 1002345-50-7
Melting Point 178°C to 183°C
Molecular Formula C27H52B2F8P2
Linear Formula C27H50P2 -+ 2HBF4
Quantity 500 mg
MDL Number MFCD12545948
Synonym dcpp
Molecular Weight (g/mol) 612.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.