Learn More
Strem, An Ascensus Company CAS# 6737-42-4. 10g. 1,3-Bis(diphenylphosphino)propane, 98% DPPP. MFCD00003050
SDP
Catalog No. 5090109883
Encompass
CAS# 6737-42-4. 10g. 1,3-Bis(diphenylphosphino)propane, 98% DPPP. MFCD00003050. Molecular Weight: 412.45. Molecular Formula: (C6H5)2PCH2CH2CH2P(C6H5)2. Color/form: white xtl. Strem# 15-0450. http://www.strem.com/catalog/v/15-0450/
Catalog No. 50-901-09883 Supplier Strem, An Ascensus Company Supplier No. 15045010G
CAS# 6737-42-4. 10g. 1,3-Bis(diphenylphosphino)propane, 98% DPPP. MFCD00003050. Molecular Weight: 412.45. Molecular Formula: (C6H5)2PCH2CH2CH2P(C6H5)2. Color/form: white xtl. Strem# 15-0450. http://www.strem.com/catalog/v/15-0450/