Learn More
Strem, An Ascensus Company CAS# 15629-92-2. 5g. 1,3-Bis(diphenylphosphino)propane nickel(II) chloride, 99%. MFCD00015318
SDP
Catalog No. 5090111219
Encompass
CAS# 15629-92-2. 5g. 1,3-Bis(diphenylphosphino)propane nickel(II) chloride, 99%. MFCD00015318. Molecular Weight: 542.08. Molecular Formula: [(C6H5)2PCH2CH2CH2P(C6H5)2]NiCl2. Color/form: red xtl. Strem# 28-0080. http://www.strem.com/catalog/v/28-0080/
Catalog No. 50-901-11219 Supplier Strem, An Ascensus Company Supplier No. 2800805G
CAS# 15629-92-2. 5g. 1,3-Bis(diphenylphosphino)propane nickel(II) chloride, 99%. MFCD00015318. Molecular Weight: 542.08. Molecular Formula: [(C6H5)2PCH2CH2CH2P(C6H5)2]NiCl2. Color/form: red xtl. Strem# 28-0080. http://www.strem.com/catalog/v/28-0080/