Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1,3-Bis(Diphenylphosphino)Propane)Palladium(II) Chloride

Catalog No. 6966762G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-667-62G 2 g
69-667-6500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 69-667-62G Supplier Sigma Aldrich Supplier No. 6966762G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1,3-Bis(Diphenylphosphino)Propane)Palladium(II) Chloride

Specifications

CAS 59831-02-6
Molecular Formula C27H26Cl2P2Pd
Linear Formula C27H26P2 -+ PdCl2
MDL Number MFCD03844773
Quantity 2 g
Synonym Dichloro[1,3-bis(diphenylphosphino)propane]palladium(II); PdCl2(dippp)
Molecular Weight (g/mol) 589.77 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.