Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(hexafluoro-alpha-hydroxyisopropyl)benzene 98.0+%, TCI America™
SDP

Catalog No. B140325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B140325G 25 g
B14035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B140325G Supplier TCI America Supplier No. B140325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 802-93-7
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.159
MDL Number MFCD00042090
InChI Key PGUIOHNOYADLMU-UHFFFAOYSA-N
Synonym 1,3-bis 2-hydroxyhexafluoroisopropyl benzene, 1,3-bis hexafluoro-2-hydroxyisopropyl benzene, 1,3-bis alpha-hydroxyhexafluoroisopropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,3-benzenedimethanol, a,a,a',a'-tetrakis trifluoromethyl-1,3-benzenedimethanol, 1,3-benzenedimethanol, .alpha.,.alpha.,.alpha.',.alpha.'-tetrakis trifluoromethyl, 1,3-bis hexafluoro-alpha-hydroxyisopropyl benzene, 1,1,1,3,3,3-hexafluoro-2-3-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,3-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, 1,3 hfab
PubChem CID 69933
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES C1=CC(=CC(=C1)C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 g
Formula Weight 410.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(hexafluoro-alpha-hydroxyisopropyl)benzene
EINECSNumber (3)-4346
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.