Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Bis(trifluoromethyl)-5-bromobenzene

Catalog No. 29015710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-015-710G 10 g
29-015-750G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-015-710G Supplier Sigma Aldrich Supplier No. 29015710G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Bis(trifluoromethyl)-5-bromobenzene

Specifications

CAS 328-70-1
Melting Point -16°C (lit.)
Density 1.699 g/mL (at 25°C (literature))
Boiling Point 154°C (lit.)
Molecular Formula C8H3BrF6
Refractive Index n20/D 1.427 (literature)
Linear Formula (CF3)2C6H3Br
MDL Number MFCD00000381
Quantity 10 g
Synonym 1-Bromo-3,5-bis(trifluoromethyl)benzene; 3,5-Bis(trifluoromethyl)bromobenzene
Molecular Weight (g/mol) 293
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.