Learn More
Accela Chembio Inc 1-(3-bromo-4-fluorophenyl)ethanol | 1g | 900175-01-1 | MFCD12147617 | 97+% | Shelf Life: 1260 Days | Light Sensitive
Supplier:  Accela Chembio Inc SY0050661G
Encompass_Preferred
1-(3-bromo-4-fluorophenyl)ethanol | 1g | 900175-01-1 | MFCD12147617 | 97+% | Shelf Life: 1260 Days | Light Sensitive
Catalog No. 50-250-669