Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(3-Bromophenoxy)acetone

Catalog No. OTV0003501G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0003501G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0003501G Supplier Sigma Aldrich Supplier No. OTV0003501G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cis-1,4-diacetoxy-2-butene is an ester
  • Ruthenium olefin metathesis catalysts having cyclic (alkyl)(amino)carbenes effectively catalyzes the cross-metathesis of cis-1,4-diacetoxy-2-butene with allylbenzene
  • Cis-1,4-diacetoxy-2-butene on pyrolysis affords 1-acetoxy-1,3-butadiene, a diacetate and an isomeric 1,2-diacetoxy-3-butene
  • Mechanism of pyrolysis has been investigated.cis-1,4-diacetoxy-2-butene may be used in the preparation of hindered diels-alder adducts
  • It may be used for the synthesis of 1,2,3,4-tetrahydro-2-vinylquinoxalines, via palladium-catalyzed tandem allylation of 1,2-phenylenediamines

Specifications

Molecular Formula C9H9BrO2
Linear Formula C9H9BrO2
MDL Number MFCD06659706
Quantity 25 mL
Molecular Weight (g/mol) 229.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.