Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(3-bromophenyl)piperazine

Catalog No. CDS014669 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014669 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014669 Supplier Sigma Aldrich Supplier No. CDS014669100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-tert-butylbenzaldehyde is an important intermediate for the synthesis of medicines, dyes, flavor and fragrance compounds
  • It is reported to be formed during the partial oxidation of 4-tert-butyltoluene by hydrogen peroxide in glacial acetic acid, catalyzed by bromide ions in combination with cobalt(ii) acetate or cerium(iii) acetate
  • Schiff base reaction between 4-tert-butylaniline and 4-tert-butylbenzaldehyde in ethanol has been carried out on-chip in the matrix assisted laser desorption ionization (maldi) chamber, the formed imine was detected in real time.4-tert-butylbenzaldehyde is suitable for use in a kinetic study to evaluate the kinetic constants (ki) for inhibition of mushroom tyrosinase by 4-substituted benzaldehydes

Specifications

CAS 31197-30-5
Molecular Formula C10H13BrN2
Linear Formula C10H13BrN2
MDL Number MFCD03412123
Quantity 25 mL
Molecular Weight (g/mol) 241.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.