Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Butadiene (ca. 15% in Toluene), TCI America™
SDP

Catalog No. B4359100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4359100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4359100ML Supplier TCI America Supplier No. B4359100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106-99-0
Molecular Formula C4H6
Molecular Weight (g/mol) 54.092
MDL Number MFCD00008659
InChI Key KAKZBPTYRLMSJV-UHFFFAOYSA-N
Synonym 1,3-butadiene, divinyl, butadiene, vinylethylene, biethylene, erythrene, bivinyl, pyrrolylene, butadieen, butadien
PubChem CID 7845
ChEBI CHEBI:39478
IUPAC Name buta-1,3-diene
SMILES C=CC=C

Specifications

Color Colorless
Quantity 100 mL
UN Number 1993
Formula Weight 54.09
Physical Form Liquid
Chemical Name or Material 1,3-Butadiene (ca. 15% in Toluene)
RTECSNumber EI9275000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.