Learn More
Medchemexpress LLC 1,3-Butanediol | 107-88-0 | 98.0% | C4H10O2 | 1 ML
SDP
Catalog No. 5000434982
Encompass_Preferred
1,3-Butanediol is an orally active ethanol dimer that provides a source of calories for human nutrition. Its administration can stimulate the biosynthesis of the ketone body, β-hydroxybutyrate (βHB). It also exhibits cerebral protective and hypoglycaemic effects.
  • Improves neurologic deficit and reduces neuronal damage in cerebral ischemia rat models.
  • Increases systemic βHB concentration in rats to levels similar to a 24-hour fast.
  • Reverses age-associated endothelium-dependent and -independent vascular dysfunction in old rats.
  • Increases plasma 3-hydroxybutyrate levels, associated with a sustained decrease in plasma glucose.
Catalog No. 50-004-34982 Supplier Medchemexpress LLC Supplier No. HY77490A10MM/1ML
1,3-Butanediol is an orally active ethanol dimer that provides a source of calories for human nutrition. Its administration can stimulate the biosynthesis of the ketone body, β-hydroxybutyrate (βHB). It also exhibits cerebral protective and hypoglycaemic effects.
  • Improves neurologic deficit and reduces neuronal damage in cerebral ischemia rat models.
  • Increases systemic βHB concentration in rats to levels similar to a 24-hour fast.
  • Reverses age-associated endothelium-dependent and -independent vascular dysfunction in old rats.
  • Increases plasma 3-hydroxybutyrate levels, associated with a sustained decrease in plasma glucose.