Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-1,3-Butanediol

Catalog No. B847851L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 g
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B847851L 500 g
B847854L 4 L
B84785100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. B847851L Supplier Sigma Aldrich Supplier No. B847851L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+/-)-1,3-Butanediol (BD) is a 1,3-diol. Its vapor pressure upto 270kPa, liquid-phase densities over a temperature range, two-phase (liquid + vapor) heat capacities, critical temperature and critical density have been determined. The obtained data was employed to derive various thermophysical properties.(+/-)-1,3-Butanediol has been used as an exogenous ketone supplement to study its effect in inducing sustained nutritional ketosis in male Sprague-Dawley rats.ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 870-46-2
Melting Point 39°C to 42°C
Boiling Point 63°C to 65°C (0.1 mmHg, lit.)
Molecular Formula C5H12N2O2
Refractive Index n20/D 1.44
Linear Formula (CH3)3COCONHNH2
MDL Number MFCD00007593
Quantity 500 g
Synonym 1,3-Butylene glycol
Molecular Weight (g/mol) 132.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.