Learn More
AdipoGen 1-3-Carboxypropyl-3 7-dmx
SDP
Supplier:  AdipoGen CDXC0311M010
Encompass_Preferred
Chemical. CAS 6493-07-8. Formula C11H14N4O4. MW 266.3. Synthetic. Major metabolite of pentoxifylline. Most effective metabolite at reducing oxygen radical production by polymorphonuclear leukocytes PMN.
Catalog No. 50-256-1387