Learn More
Sigma Aldrich 1-(3-CHLORO-4-FLUOROPHENYL)-4-(4-CHLOROPHENYL)-1H-PYRAZOL-5-AMINE
Catalog No. JRD06161G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
JRD06161G 5 g
1 options
Catalog No. JRD06161G Supplier Sigma Aldrich Supplier No. JRD06161G
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
  • Ethyl (4-fluorobenzoyl)acetate on condensation with benzofurazan oxide yields 2-(carboethoxy)-3-(4α-fluoro)phenylquinoxaline1,4-dioxide.reactant used as a precursor in:
  • Condensation reactions with diamines via c-c bond cleavage for synthesis of benzimidazoles and perimidines for possible use as antimalarial treatments
  • Base-promoted domino michael addition / cyclization / elimination reactions for synthesis of hydroxybenzophenones
  • Oxidative cross-coupling with indoles via dioxygen activation
  • Cyclization of keto esters for synthesis of pyrones
  • Lewis base catalyzed hydrosilylation for synthesis of α-acetoxy α-amino acid derivatives
  • Conia-ene reactions for synthesis of methylenecyclopentane derivatives
Specifications
Quantity 5 g