Learn More
Sigma Aldrich 1-(3-Chlorophenyl)cyclopentanamine hydrochloride
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1-bromo-4-methylpentane can be synthesized by treating 4-methyl-1-pentanol with phosphorous tribromide. the conformational analysis of its liquid-state and solid-state IR and raman spectra and showed the presence of a mixture of pc,pH, and pαh conformers.1-bromo-4-methylpentane may be used in the synthesis of:
- Diethyl 2-(4α-methylpentyl)malonate
- 1,3-bis(4-(isopentyloxy)phenyl)urea
- 1,3-bis(4-(isopentyloxy)phenyl)thiourea
- 13-methyl-1-[(tetrahydropyran-2-yl)oxy]tetradec-8-yne
- 15-methyl-1-[(tetrahydropyran-2-yl)oxy]hexadec-10-yne
- 3-[2-isohexyloxy-3-(hydroxymethyl]-5-phenyl-2-isohexyloxybenzyl alcohol
Specifications
Specifications
| CAS | 1215574-59-6 |
| Molecular Formula | C11H15Cl2N |
| Linear Formula | C11H15Cl2N |
| MDL Number | MFCD15142053 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 232.15 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.