Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(3-Chlorophenyl)piperazine hydrochloride

Catalog No. 12518025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-518-025G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-518-025G Supplier Sigma Aldrich Supplier No. 12518025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-(3-Chlorophenyl)piperazine hydrochloride

Specifications

CAS 65369-76-8
Melting Point 210°C to 214°C (dec.) (lit.)
Molecular Formula C10H13ClN2 -+ xHCl
Linear Formula C10H13ClN2 -+ xHCl
MDL Number MFCD00039032
Quantity 250 mg
Synonym m-CPP
Molecular Weight (g/mol) 196.68 (free base basis)
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.