Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Chlorophenyl)piperazine hydrochloride, 97%, Thermo Scientific™

Catalog No. AAH5989606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5989606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5989606 Supplier Thermo Scientific Chemicals Supplier No. H5989606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65369-76-8
Molecular Formula C10H14Cl2N2
Molecular Weight (g/mol) 233.136
MDL Number MFCD00039032
InChI Key MHXPYWFZULXYHT-UHFFFAOYSA-N
Synonym 1-3-chlorophenyl piperazine hydrochloride, m-cpp hydrochloride, 1-3-chlorophenyl piperazine monohydrochloride, unii-7im2gs7ilv, 1-3-chlorophenyl piperazinium chloride, piperazine, 1-3-chlorophenyl-, monohydrochloride, 1-m-chlorophenyl piperazine hcl, m-chlorophenylpiperazine hydrochloride, 7im2gs7ilv, 1-m-chlorophenyl piperazine hydrochloride
PubChem CID 25681
IUPAC Name 1-(3-chlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC(=CC=C2)Cl.Cl

Specifications

Melting Point 210°C to 214°C (decomposition)
Color Brown
Quantity 5 g
Formula Weight 196.68 (Anhydrous)
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(3-Chlorophenyl)piperazine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 265-718-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.