Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Chlorophenyl)piperazine monohydrochloride, 97%, Thermo Scientific Chemicals

Catalog No. AAA1405709
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1405709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1405709 Supplier Thermo Scientific Chemicals Supplier No. A1405709
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13078-15-4
Molecular Formula C10H14Cl2N2
Molecular Weight (g/mol) 233.14
MDL Number MFCD00039032,MFCD00012764
InChI Key MHXPYWFZULXYHT-UHFFFAOYSA-N
Synonym 1-3-chlorophenyl piperazine hydrochloride, m-cpp hydrochloride, 1-3-chlorophenyl piperazine monohydrochloride, unii-7im2gs7ilv, 1-3-chlorophenyl piperazinium chloride, piperazine, 1-3-chlorophenyl-, monohydrochloride, 1-m-chlorophenyl piperazine hcl, m-chlorophenylpiperazine hydrochloride, 7im2gs7ilv, 1-m-chlorophenyl piperazine hydrochloride
PubChem CID 25681
IUPAC Name hydrogen 1-(3-chlorophenyl)piperazine chloride
SMILES [H+].[Cl-].ClC1=CC=CC(=C1)N1CCNCC1

Specifications

Melting Point 214°C (decomposition)
Quantity 10 g
UN Number UN2811
Beilstein 4768691
Sensitivity Hygroscopic
Formula Weight 233.14
Percent Purity 97%
Chemical Name or Material 1-(3-Chlorophenyl)piperazine monohydrochloride
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 235-976-9
RTECSNumber TL2833500
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.