Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Cyanopropyl)pyrrolidine 95.0+%, TCI America™
SDP

Catalog No. C123125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C123125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C123125ML Supplier TCI America Supplier No. C123125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35543-25-0
Molecular Formula C8H14N2
Molecular Weight (g/mol) 138.214
MDL Number MFCD00003183
InChI Key OSDDDHPYSNZBPF-UHFFFAOYSA-N
Synonym 1-pyrrolidinebutanenitrile, 4-pyrrolidin-1-yl butanenitrile, 1-pyrrolidinebutyronitrile, 1-3-cyanopropyl pyrrolidine, 4-1-pyrrolidine butyronitrile, 1-pyrrolidinobutyronitrile, 4-pyrrolidin-1-yl-butyronitrile, 4-pyrrolidinylbutanenitrile, 4-pyrrolidin-1-ylbutyronitrile, 4-pyrrolidinobutyronitrile
PubChem CID 118838
IUPAC Name 4-pyrrolidin-1-ylbutanenitrile
SMILES C1CCN(C1)CCCC#N

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 mL
Formula Weight 138.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Cyanopropyl)pyrrolidine
RTECSNumber UY0270000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.