Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Cyclohexanedione 98.0+%, TCI America™
SDP

Catalog No. A55815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A55815G Supplier TCI America Supplier No. A55815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 504-02-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001585
InChI Key HJSLFCCWAKVHIW-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
PubChem CID 10434
ChEBI CHEBI:17766
IUPAC Name cyclohexane-1,3-dione
SMILES C1CC(=O)CC(=O)C1

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 5 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Cyclohexanedione [for HPLC Labeling]
EINECSNumber (9)-0519
RTECSNumber GV0350000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.