Learn More
Medchemexpress LLC 1,3-Cyclopentanedicarboxylic acid, 1-amino-, (1R,3S)-rel- | 67684-64-4 | MFCD00153759 | 98.0% | 173.168 | 5 MG
SDP
Catalog No. 5000384783
Encompass_Preferred
trans-ACPD is a metabotropic receptor agonist that induces calcium mobilization and an inward current in cultured cerebellar Purkinje neurons. It is intended for research use only.
  • A metabotropic receptor agonist.
  • Produces calcium mobilization.
  • Generates an inward current in cultured cerebellar Purkinje neurons.
Catalog No. 50-003-84783 Supplier Medchemexpress LLC Supplier No. HY194345MG
trans-ACPD is a metabotropic receptor agonist that induces calcium mobilization and an inward current in cultured cerebellar Purkinje neurons. It is intended for research use only.
  • A metabotropic receptor agonist.
  • Produces calcium mobilization.
  • Generates an inward current in cultured cerebellar Purkinje neurons.