Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Di(4-pyridyl)propane 97.0+%, TCI America™
SDP

Catalog No. D093825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D093825G 25 g
D0938500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D093825G Supplier TCI America Supplier No. D093825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17252-51-6
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD00038046
InChI Key OGNCVVRIKNGJHQ-UHFFFAOYSA-N
Synonym 4,4'-trimethylenedipyridine, 1,3-di 4-pyridyl propane, 1,3-bis 4-pyridyl propane, pyridine, 4,4'-1,3-propanediyl bis, 4,4'-propane-1,3-diyldipyridine, 4-3-pyridin-4-ylpropyl pyridine, 4-3-4-pyridyl propyl pyridine, 4-3-pyridin-4-yl propyl pyridine, pyridine, 4,4'-trimethylenedi, 4,4'-propane-1,3-diyl dipyridine
PubChem CID 87019
IUPAC Name 4-(3-pyridin-4-ylpropyl)pyridine
SMILES C1=CN=CC=C1CCCC2=CC=NC=C2

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 162°C
Quantity 25 g
Formula Weight 198.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di(4-pyridyl)propane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.