Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Di-tert-butylbenzene, 99%

Catalog No. AC448890010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448890010 1 g Glass bottle
AC448890050 5 g Glass bottle
AC448890250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC448890010 Supplier Thermo Scientific Chemicals Supplier No. 448890010
Add to Cart

CAS: 1014-60-4 | C14H22 | 190.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1014-60-4
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00008830
InChI Key ILNDSSCEZZFNGE-UHFFFAOYSA-N
Synonym 1,3-di-tert-butylbenzene, benzene, 1,3-bis 1,1-dimethylethyl, benzene, m-di-tert-butyl, m-di-tert-butylbenzene, 1,3-di-t-butylbenzene, 1,3-ditert-butyl-benzene, 1,3-ditertiarybutylbenzene, 1,3-di-tert-butyl-benzene, acmc-1bo58, 1,3-bis tert-butyl benzene
PubChem CID 136810
IUPAC Name 1,3-ditert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC=C1)C(C)(C)C

Specifications

Melting Point 10.0°C to 11.0°C
Density 0.8600g/mL
Boiling Point 106.0°C to 107.0°C (18.0 mmHg)
Flash Point 84°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CC6H4C(CH3)3
Quantity 1 g
Specific Gravity 0.86
Formula Weight 190.33
Percent Purity 99%
Chemical Name or Material 1,3-Di-tert-butylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.