Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diacetylbenzene 98.0+%, TCI America™
SDP

Catalog No. D26175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26175G 5 g
D26171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26175G Supplier TCI America Supplier No. D26175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6781-42-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008740
InChI Key VCHOFVSNWYPAEF-UHFFFAOYSA-N
Synonym 1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene
PubChem CID 23229
IUPAC Name 1-(3-acetylphenyl)ethan-1-one
SMILES CC(=O)C1=CC(=CC=C1)C(C)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,3-Diacetylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.