Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diallylurea 99%, Thermo Scientific™

Catalog No. AC202880050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC202880050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC202880050 Supplier Thermo Scientific Chemicals Supplier No. 202880050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1801-72-5
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
InChI Key QRWVOJLTHSRPOA-UHFFFAOYSA-N
Synonym 1,3-diallylurea, n,n'-diallylurea, urea, n,n'-di-2-propenyl, urea, 1,3-diallyl, unii-lxm0xy469i, diallyl urea, lxm0xy469i, urea,n,n'-di-2-propen-1-yl, diallylurea, sinapoline
PubChem CID 74537
IUPAC Name 1,3-bis(prop-2-enyl)urea
SMILES C=CCNC(=O)NCC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 90°C to 93°C
Color White to Yellow
Infrared Spectrum Authentic
Linear Formula H2C=CHCH2NHCONHCH2CH=CH2
Quantity 5 g
Beilstein 04, 209
Formula Weight 140.18
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 3-Diallylurea, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.