Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diallylurea 98.0+%, TCI America™
SDP

Catalog No. D51701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51701G 1 g
D51705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51701G Supplier TCI America Supplier No. D51701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1801-72-5
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00008639
InChI Key QRWVOJLTHSRPOA-UHFFFAOYSA-N
Synonym 1,3-diallylurea, n,n'-diallylurea, urea, n,n'-di-2-propenyl, urea, 1,3-diallyl, unii-lxm0xy469i, diallyl urea, lxm0xy469i, urea,n,n'-di-2-propen-1-yl, diallylurea, sinapoline
PubChem CID 74537
IUPAC Name 1,3-bis(prop-2-enyl)urea
SMILES C=CCNC(=O)NCC=C

Specifications

Melting Point 98°C
Color White
Boiling Point 56°C
Quantity 1 g
Formula Weight 140.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diallylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.