Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic acid, 98+%, Thermo Scientific™

Catalog No. AAA1746322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1746322 100 g
AAA1746314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1746322 Supplier Thermo Scientific Chemicals Supplier No. A1746322

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3148-72-9
Molecular Formula C11H18N2O9
Molecular Weight (g/mol) 322.27
MDL Number MFCD00004288
InChI Key WYMDDFRYORANCC-UHFFFAOYSA-N
Synonym dhpta, dpta, 1,3-diaminopropanol tetraacetic acid, diaminohydroxypropane tetraacetic acid, 1,3-diamino-2-propanol-n,n,n',n'-tetraacetic acid, diaminopropanol tetraacetic acid, unii-949b9zmo7m, 1,3-diamino-2-hydroxypropane-n,n,n',n'-tetraacetic acid, 2-hydroxy-1,3-propylenediaminetetraacetic acid, diaminopropanol tetra acetic acid
PubChem CID 18465
IUPAC Name 2-[[3-[bis(carboxymethyl)amino]-2-hydroxypropyl]-(carboxymethyl)amino]acetic acid
SMILES C(C(CN(CC(=O)O)CC(=O)O)O)N(CC(=O)O)CC(=O)O

Specifications

Melting Point ∼195°C (decomposition)
Quantity 100 g
Beilstein 1716800
Formula Weight 322.27
Percent Purity ≥98%
Chemical Name or Material 1,3-Diamino-2-propanol-N,N,N',N'-tetraacetic acid
Hazard Category H312-H315-H319-H335
Hazard Statement GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-575-6
RTECSNumber AI2930000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.