Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diaminopropane-N,N'-diacetic Acid 98.0+%, TCI America™
SDP

Catalog No. P111525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P111525G 25 g
P11155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P111525G Supplier TCI America Supplier No. P111525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112041-05-1
Molecular Formula C7H14N2O4
Molecular Weight (g/mol) 190.199
MDL Number MFCD00142534
InChI Key RKPHYUBLENEQNT-UHFFFAOYSA-N
PubChem CID 414255
IUPAC Name 2-[3-(carboxymethylamino)propylamino]acetic acid
SMILES C(CNCC(=O)O)CNCC(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Synonym 1,3-Propylenediamine-N,N′-diacetic Acid
Formula Weight 190.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diaminopropane-N,N′-diacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.