Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibenzylidene-2-cyclohexanone 98.0+%, TCI America™
SDP

Catalog No. D11941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11941G Supplier TCI America Supplier No. D11941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 897-78-9
Molecular Formula C20H18O
Molecular Weight (g/mol) 274.363
MDL Number MFCD00019516
InChI Key CTKKGXDAWIAYSA-JSAVKQRWSA-N
Synonym 2,6-dibenzylidenecyclohexanone, cyclohexanone, 2,6-dibenzylidene, 1,3-dibenzylidene-2-cyclohexanone, 2e,6e-2,6-dibenzylidenecyclohexanone, 2,6-bis benzylidene cyclohexanone, 2e,6e-2,6-dibenzylidenecyclohexan-1-one, cyclohexanone, 2,6-bis phenylmethylene, cyclohexanone,6-dibenzylidene, 2e,6e-2,6-bis phenylmethylidene cyclohexan-1-one, 2,6-bisbenzylidenecyclohexanone
PubChem CID 1550330
IUPAC Name (2E,6E)-2,6-dibenzylidenecyclohexan-1-one
SMILES C1CC(=CC2=CC=CC=C2)C(=O)C(=CC3=CC=CC=C3)C1

Specifications

Melting Point 117°C
Color Yellow
Quantity 1 g
UN Number 3077
Formula Weight 274.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibenzylidene-2-cyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.