Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-Dibromo-2,2-dimethoxypropane | 22094-18-4 | 261.94 | 50 G
SDP

Catalog No. 5000445479
Encompass_Preferred

1,3-Dibromo-2,2-dimethoxypropane is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Solid appearance
  • White to off-white color
  • For research use only, not intended for human use or for sale to patients
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Purity: 99.98%
  • Molecular weight: 261.94

Catalog No. 50-004-45479 Supplier Medchemexpress LLC Supplier No. HYW00501250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1,3-Dibromo-2,2-dimethoxypropane is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Solid appearance
  • White to off-white color
  • For research use only, not intended for human use or for sale to patients
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Purity: 99.98%
  • Molecular weight: 261.94

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.