Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-dibromo-2-propanol | 96-21-9 | MFCD00000216 | 99.7% | 217.89 g/mol | C3H6Br2O | 50 G
SDP

Catalog No. 5000304053
Encompass_Preferred

1,3-Dibromo-2-propanol is a dibrominated propanol used as a biochemical reagent and as an intermediate in organic synthesis. The product is supplied with a certificate of analysis and a safety data sheet for research use; handle according to standard laboratory safety procedures.

  • High purity: 99.7%.
  • Molecular weight 217.89 g/mol.
  • Chemical formula C3H6Br2O.
  • Physical form: liquid.
  • Common applications include use as a biochemical reagent and a synthetic intermediate.
  • Storage guidance: store at room temperature; in solvent, -80°C (2 years) or -20°C (1 year).

Catalog No. 50-003-04053 Supplier Medchemexpress LLC Supplier No. HYW01409250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dibromo-2-propanol is a dibrominated propanol used as a biochemical reagent and as an intermediate in organic synthesis. The product is supplied with a certificate of analysis and a safety data sheet for research use; handle according to standard laboratory safety procedures.

  • High purity: 99.7%.
  • Molecular weight 217.89 g/mol.
  • Chemical formula C3H6Br2O.
  • Physical form: liquid.
  • Common applications include use as a biochemical reagent and a synthetic intermediate.
  • Storage guidance: store at room temperature; in solvent, -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.