Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-dibromo-2-propanol | 96-21-9 | MFCD00000216 | 99.7% | 217.89 g/mol | C3H6Br2O | 500 G
SDP

Catalog No. 5000304004
Encompass_Preferred

1,3-Dibromo-2-propanol is a dibrominated secondary alcohol used as a biochemical reagent and intermediate in organic synthesis. It is used in laboratory research for synthesis, derivatization, and as a building block in chemical reactions, and is supplied with analytical documentation.

  • High purity (≈99.7%).
  • Molecular formula C3H6Br2O; molecular weight 217.89 g/mol.
  • Suitable as a synthetic intermediate and reagent for organic transformations.
  • Available in multiple pack sizes including 500 g.
  • Certificate of analysis and product data sheet available.
  • Store at room temperature; solutions may require cold storage.

Catalog No. 50-003-04004 Supplier Medchemexpress LLC Supplier No. HYW014092500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dibromo-2-propanol is a dibrominated secondary alcohol used as a biochemical reagent and intermediate in organic synthesis. It is used in laboratory research for synthesis, derivatization, and as a building block in chemical reactions, and is supplied with analytical documentation.

  • High purity (≈99.7%).
  • Molecular formula C3H6Br2O; molecular weight 217.89 g/mol.
  • Suitable as a synthetic intermediate and reagent for organic transformations.
  • Available in multiple pack sizes including 500 g.
  • Certificate of analysis and product data sheet available.
  • Store at room temperature; solutions may require cold storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.