Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-5-fluoro-2-iodobenzene 98.0+%, TCI America™
SDP

Catalog No. D35825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35825G Supplier TCI America Supplier No. D35825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62720-29-0
Molecular Formula C6H2Br2FI
Molecular Weight (g/mol) 379.79
MDL Number MFCD00042232
InChI Key LIWKANDEJFABTQ-UHFFFAOYSA-N
Synonym 2,6-dibromo-4-fluoroiodobenzene, benzene, 1,3-dibromo-5-fluoro-2-iodo, 1,3-dibromo-5-fluoro-2-iodo-benzene, pubchem3449, acmc-1b2cd, ksc615o5d, 2.6-dibromo-4-fluoro iodobenzene, 2,6-dibromo-4-fluoro-1-iodobenzene, benzene,1,3-dibromo-5-fluoro-2-iodo
PubChem CID 2724606
IUPAC Name 1,3-dibromo-5-fluoro-2-iodobenzene
SMILES FC1=CC(Br)=C(I)C(Br)=C1

Specifications

Melting Point 134°C
Quantity 5 g
Formula Weight 379.79
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromo-5-fluoro-2-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.