Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromoadamantane 97.0+%, TCI America™
SDP

Catalog No. D24805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24805G 5 g
D248025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24805G Supplier TCI America Supplier No. D24805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 876-53-9
Molecular Formula C10H14Br2
Molecular Weight (g/mol) 294.03
MDL Number MFCD00077218
InChI Key HLWZKLMEOVIWRK-UHFFFAOYSA-N
PubChem CID 265790
IUPAC Name 1,3-dibromoadamantane
SMILES C1C2CC3(CC1CC(C2)(C3)Br)Br

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 5 g
Formula Weight 294.03
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dibromoadamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.