Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromobenzene, 97%

Catalog No. AC186640100
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186640100 10 mL Glass bottle
AC186640500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186640100 Supplier Thermo Scientific Chemicals Supplier No. 186640100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 108-36-1 | C6H4Br2 | 235.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-36-1
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000078
InChI Key JSRLURSZEMLAFO-UHFFFAOYSA-N
Synonym m-dibromobenzene, benzene, 1,3-dibromo, benzene, m-dibromo, unii-74ef6kh8tc, 1,3-dibromo-benzene, 74ef6kh8tc, 1,3-dibromo benzene, 3,5-dibromobenzene, 1,3-dibrormobenzene, 1, 3-dibromobenzene
PubChem CID 7927
ChEBI CHEBI:37151
IUPAC Name 1,3-dibromobenzene
SMILES BrC1=CC(Br)=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -7°C
Color Colorless to Yellow
Density 1.9500g/mL
Boiling Point 218°C to 219°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.6070 to 1.609
Linear Formula C6H4Br2
Quantity 10 mL
Beilstein 05, 211
Specific Gravity 1.95
Solubility Information Solubility in water: insoluble
Formula Weight 235.91
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 3-Dibromobenzene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-574-2
RTECSNumber CZ1790000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.