Learn More
Medchemexpress LLC 1,3-dibromopropan-2-ol | 96-21-9 | MFCD00000216 | 99.7% | 217.89 | C3H6Br2O | 5 G
SDP
Catalog No. 5000304093
Encompass_Preferred
1,3-dibromopropan-2-ol is an aliphatic vicinal dibromohydrin used as a biochemical reagent and intermediate in organic synthesis. It is typically a colorless to pale-yellow liquid with molecular formula C3H6Br2O and molecular weight 217.89 g·mol-1. Reported purity is about 99.7%; available in laboratory pack sizes. Storage is at room temperature for short term, with colder conditions recommended for long-term storage in solvent.
Catalog No. 50-003-04093 Supplier Medchemexpress LLC Supplier No. HYW0140925G
1,3-dibromopropan-2-ol is an aliphatic vicinal dibromohydrin used as a biochemical reagent and intermediate in organic synthesis. It is typically a colorless to pale-yellow liquid with molecular formula C3H6Br2O and molecular weight 217.89 g·mol-1. Reported purity is about 99.7%; available in laboratory pack sizes. Storage is at room temperature for short term, with colder conditions recommended for long-term storage in solvent.