Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dibromopropane

Catalog No. 1259031L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-590-31L 1 L
12-590-350ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-590-31L Supplier Sigma Aldrich Supplier No. 1259031L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Dibromopropane

Specifications

CAS 109-64-8
Melting Point -34°C
Density 1.989 g/mL at 25°C
Boiling Point 167°C
Molecular Formula C3H6Br2
Refractive Index n20/D 1.524
Linear Formula Br(CH2)3Br
MDL Number MFCD00000255
Quantity 1 L
Synonym Trimethylene dibromide
Molecular Weight (g/mol) 201.89 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.