Learn More
Medchemexpress LLC 1,3-dibromopropane (trimethylene dibromide) | 109-64-8 | MFCD00000255 | 5g
SDP
Catalog No. 5000266584
Encompass_Preferred
1 3-Dibromopropane (Trimethylene dibromide) is a standard substrate 1 3-Dibromopropane can be used for haloalkane dehalogenase DmmA assays[1]
Catalog No. 50-002-66584 Supplier Medchemexpress LLC Supplier No. HYY18065G
1 3-Dibromopropane (Trimethylene dibromide) is a standard substrate 1 3-Dibromopropane can be used for haloalkane dehalogenase DmmA assays[1]