Learn More
Cayman Chemical 1,3-Dicaffeoylquinic Acid, Purity: ≥98%, CAS: 30964-13-7, 50 mg
SDP
Catalog No. 502932475
Encompass_Preferred
A polyphenol; decreases the expression and protein levels of iNOS at 10 µM in HCASMCs treated with a cytokine mixture; decreases protein levels of tyrosinase and MITF in B16F1 cells; inhibits melanogenesis in B16F1 cells and decreases tyrosinase activity in cell lysates at 25 µM; reduces triglyceride levels in sodium oleate-induced hyperlipidemic HepG2 cells at 20 and 40 µM
Catalog No. 50-293-2475 Supplier Cayman Chemical Supplier No. 2502250 MG
A polyphenol; decreases the expression and protein levels of iNOS at 10 µM in HCASMCs treated with a cytokine mixture; decreases protein levels of tyrosinase and MITF in B16F1 cells; inhibits melanogenesis in B16F1 cells and decreases tyrosinase activity in cell lysates at 25 µM; reduces triglyceride levels in sodium oleate-induced hyperlipidemic HepG2 cells at 20 and 40 µM