Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-1,1,3,3-tetramethyldisiloxane 97.0+%, TCI America™
SDP

Catalog No. D23345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23345G 5 g
D233425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23345G Supplier TCI America Supplier No. D23345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2401-73-2
Molecular Formula C4H12Cl2OSi2
Molecular Weight (g/mol) 203.209
MDL Number MFCD00018088
InChI Key DMEXFOUCEOWRGD-UHFFFAOYSA-N
Synonym 1,3-dichloro-1,1,3,3-tetramethyldisiloxane, 1,3-dichlorotetramethyldisiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetramethyl, chloro chlorodimethylsilyl oxy dimethylsilane, 1,1,3,3-tetramethyl-1,3-dichlorodisiloxane, acmc-209g7w, oxybis chlorodimethylsilane, sym-dichlorotetramethyldisiloxane, dmexfouceowrgd-uhfffaoysa, 1,3-dichlorotetramethyl disiloxane
PubChem CID 75461
IUPAC Name chloro-[chloro(dimethyl)silyl]oxy-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)Cl)Cl

Specifications

Color Yellow
Boiling Point 138°C
Quantity 5 g
UN Number 2986
Formula Weight 203.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetramethyldisiloxane
EINECSNumber (7)-0472
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.