Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-2-methoxy-5-nitrobenzene, 99%, Thermo Scientific™

Catalog No. AC271022500
Change view
Click to view available options
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC271022500 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC271022500 Supplier Thermo Scientific Chemicals Supplier No. 271022500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 17742-69-7
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H5Cl2NO3
Quantity 250g
Synonym 2,6-dichloro-4-nitroanisole, 2,6-dichloro-4-nitro-anisole, 2,6-dichloro-4-nitrophenyl methyl ether, benzene, 1,3-dichloro-2-methoxy-5-nitro, pubchem2687, acmc-1c7kz, gjyvjkpfycknec-uhfffaoysa, 1,3-dichloro-2-methoxy-5-nitrobe, 3,5-dichloro-4-methoxynitrobenzene, 1,3-dichloro-2-methoxy-5-nitro-benzene
InChI Key GJYVJKPFYCKNEC-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1Cl)[N+](=O)[O-])Cl
IUPAC Name 1,3-dichloro-2-methoxy-5-nitrobenzene
Molecular Weight (g/mol) 222.021
PubChem CID 309716
Formula Weight 222.03
Percent Purity ≥98.5%
Chemical Name or Material 1, 3-Dichloro-2-methoxy-5-nitrobenzene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.