Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-2-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. D344925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D344925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D344925G Supplier TCI America Supplier No. D344925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 601-88-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00456795
InChI Key VITSNECNFNNVQB-UHFFFAOYSA-N
Synonym 2,6-Dichloronitrobenzene
PubChem CID 11759
IUPAC Name 1,3-dichloro-2-nitrobenzene
SMILES [O-][N+](=O)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 192.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dichloro-2-nitrobenzene
EINECSNumber (3)-0455
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.