Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dichloro-2-propanol

Catalog No. 501859591
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-9591 100 g
50-185-9592 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-9591 Supplier Sigma Aldrich Supplier No. 184489100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Dichloro-2-propanol
TRUSTED_SUSTAINABILITY

Specifications

CAS 96-23-1
Density 1.363 g/mL at 20°C, 1.351 g/mL (at 25°C (literature))
Molecular Formula C3H6Cl2O
Linear Formula (ClCH2)2 CHOH
MDL Number MFCD00000951
Melting Point -4°C (lit.)
Boiling Point 174.3°C (lit.)
Synonym Glycerol-alpha,gamma-dichlorohydrin
Molecular Weight (g/mol) 128.99
Quantity 100 g
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.