Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dicyclohexylimidazolium Tetrafluoroborate Salt

Catalog No. 6661815G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-618-15G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-618-15G Supplier Sigma Aldrich Supplier No. 6661815G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Dicyclohexylimidazolium Tetrafluoroborate Salt

Specifications

CAS 274257-37-3
Molecular Formula C15H33NO3
Linear Formula C15H33NO3
MDL Number MFCD09265058
Quantity 10 g
Molecular Weight (g/mol) 275.43 g/mol
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.