Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3-dicyclohexylurea | 2387-23-7 | MFCD00003829 | 99.7% | 224.34 | C13H24N2O | 5 MG
SDP

Catalog No. 5000303995
Encompass_Preferred

1,3-Dicyclohexylurea is supplied as a white solid analytical standard intended for research and analytical applications. The compound (C13H24N2O, MW 224.34) is provided at high assay purity and is packaged to reduce exposure to light and moisture; store sealed at 4°C to preserve stability.

  • High purity (99.66%) suitable for analytical standards and assay verification.
  • White solid appearance for straightforward identification.
  • Molecular weight 224.34 and formula C13H24N2O for unambiguous reference.
  • Brown glass packaging protects from light exposure.
  • Storage at 4°C maintains compound stability.
  • Suitable as a reference material for analytical and research workflows.

Catalog No. 50-003-03995 Supplier Medchemexpress LLC Supplier No. HYW013989R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dicyclohexylurea is supplied as a white solid analytical standard intended for research and analytical applications. The compound (C13H24N2O, MW 224.34) is provided at high assay purity and is packaged to reduce exposure to light and moisture; store sealed at 4°C to preserve stability.

  • High purity (99.66%) suitable for analytical standards and assay verification.
  • White solid appearance for straightforward identification.
  • Molecular weight 224.34 and formula C13H24N2O for unambiguous reference.
  • Brown glass packaging protects from light exposure.
  • Storage at 4°C maintains compound stability.
  • Suitable as a reference material for analytical and research workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.