Learn More
Medchemexpress LLC 1,3-dicyclohexylurea | 2387-23-7 | MFCD00003829 | 99.9% | 224.34 g/mol | C13H24N2O | 100 MG
SDP
Catalog No. 5000303879
Encompass_Preferred
1,3-Dicyclohexylurea (DCU) is a small-molecule soluble epoxide hydrolase (sEH) inhibitor provided as a high-purity biochemical reagent for enzymology and pharmacology research. It is supplied in both solid and solution formats for in vitro assays and formulation studies, with documented solubility and storage recommendations to support reproducible handling.
Catalog No. 50-003-03879 Supplier Medchemexpress LLC Supplier No. HYW013989100MG
1,3-Dicyclohexylurea (DCU) is a small-molecule soluble epoxide hydrolase (sEH) inhibitor provided as a high-purity biochemical reagent for enzymology and pharmacology research. It is supplied in both solid and solution formats for in vitro assays and formulation studies, with documented solubility and storage recommendations to support reproducible handling.