Learn More
Cayman Chemical 1,3-Didocosahexaenoyl Glycerol, Purity: ≥98%, CAS: 140670-42-4, 5 mg
SDP
Catalog No. 502939623
Encompass_Preferred
A diacylglycerol; has been used in the synthesis of triacylglycerols that contain DHA at their sn-1 and sn-3 positions
Catalog No. 50-293-9623 Supplier Cayman Chemical Supplier No. 270115 MG
A diacylglycerol; has been used in the synthesis of triacylglycerols that contain DHA at their sn-1 and sn-3 positions